Learn More
TraceCERT™ Molybdenum Standard for ICP-MS, 1 mg/L Mo in Water (with 2 Ammonium Hydroxide), MilliporeSigma™ Supelco™
Supplier:  MilliporeSigma Supelco 04488100ML
Catalog No. 11-101-6860
Add to Cart
Description
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Specifications
Specifications
1336-21-6
1 mg/L Mo aq. soln. (with 2% ammonium hydroxide)
100 mL
Certified Reference Material
1 mg/L Mo aq. soln., prepared with high purity Ammonium molybdatde, Ammonium hydroxide solution and water
TraceCERT™ Analytical Standard
TraceCERT
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC