Learn More
TraceCERT™ Lithium Standard for ICP-MS, 1 mg/L Li in Nitric Acid, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 39185100ML
Catalog No. 11-101-6761
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Specifications
Specifications
7697-37-2
1 mg/L Li in nitric acid
100 mL
Certified Reference Material
1 mg/L Li in 2% nitric acid, prepared with high purity Li2 CO3 and HNO3
TraceCERT
TraceCERT
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC