Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Gasoline, premium unleaded, refinery grade, 20,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 47516U
Description
Environmental and PetroleumSpecifications
8032-32-4 | |
Petrol; Gasoline; Petroleum benzin | |
20,000 µg/mL in methanol | |
1 mL | |
TraceCERT |
MFCD00081849 | |
TraceCERT™ Analytical Standard | |
Ampule | |
9°C | |
Certified Reference Material |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC