Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Atrazine Solution, 1,000 μg/mL in methyl tert-butyl ether, Certified Reference Material, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 48187
Description
Agriculture and EnvironmentalSpecifications
1912-24-9 | |
MFCD00041810 | |
TraceCERT™ Analytical Standard | |
Ampule | |
-10°C | |
Certified Reference Material |
C8H14 ClN5 | |
215.68 | |
1,000 µg/mL in methyl tert -butyl ether | |
1 mL | |
TraceCERT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC