Learn More
Andwin Scientific TPP Cell Scrapers, 38 cm, 100/cs
Supplier: Andwin Scientific TP99004
TPP Cell Scrapers have a pivoting head that enables the blade angle to be changed effortlessly with a slight twist of the fingers while still in contact with the surface being scraped. This allows for easy and thorough cell retrieval from vessels of all shapes and sizes without having to constantly re-position the scraper or culture vessel. The TPP exclusive raised knobs on the handle allow for a slip free and secure grip when wearing gloves.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.