Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Tote Tray and Shelf Storage Cabinet
SCI_ED

Ideal for storing projects and larger objects. The doors have three-point locking handles for secure storage

$3559.00 - $3862.00

Specifications

Certifications/Compliance SEFA, Mas Certified Green
Finish Chemical Resistant, Earth-friendly UV
For Use With (Application) Storage/Science classrooms
Includes Locks
Warranty Limited lifetime
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
S37974
SCI_ED
View Documents
Diversified Spaces™
3514822K
Oak
Each for $3,559.00
Add to Cart
 
S37975
SCI_ED
View Documents
Diversified Spaces™
3514822M
Maple
Each for $3,862.00
Add to Cart
 
Description

Description

  • Finished with a chemical resistant, earth-friendly UV finish and rubber base molding
  • 2 adjustable and one fixed shelf
  • Holds up to 24 tote trays (tote trays are included)
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Features two hinged oak doors with three-point locking handles
Specifications

Specifications

SEFA, Mas Certified Green
Storage/Science classrooms
Limited lifetime
Tall Storage Cabinets with Doors
Chemical Resistant, Earth-friendly UV
Locks
48 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit