Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - FT3 (Free Triiodothyronine)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$398.00
Specifications
For Use With (Application) | Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |
---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
21-279168
|
Tosoh Bioscience
020386 |
12 x 1 mL | FT3 Calibrator Set |
Each for $398.00
|
|
||||
21-280-003
|
Tosoh Bioscience
025286 |
5 trays of 20 cups | FT3 Test Cup |
Each for $1,043.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time |