Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - BMG (Beta-2-Microglobulin)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Products 2
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
21-279-34
View Documents
Tosoh Bioscience
020359
1 mL BMG (beta-2-Microglobulin) Calibrator Set
Each for $387.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
21-279-60
View Documents
Tosoh Bioscience
020559
10 mL BMG (beta-2-Microglobulin) Sample Diluting Concentrate
Each for $136.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Safety and Handling
Safety and Handling
ShelfLife : 1 Year