Learn More
Tosoh Bioscience AIA-PACK™ Test Cups – B12 Vitamin
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
$191.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
21-279170
|
Tosoh Bioscience
020706 |
Vitamin B12 Pretreatment Set |
Each for $111.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-279-35
|
Tosoh Bioscience
020393 |
Vitamin B12 Calibrator Set |
Each for $409.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-279-63
|
Tosoh Bioscience
020593 |
Vitamin B12 Sample Diluting Solution |
Each for $191.00
|
|
|||||
21-279-9
|
Tosoh Bioscience
020293 |
Vitamin B12 Assay Test Cup |
Each for $2,320.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
- Saves hands-on time for operators and reduces waste
- No pre-mixing or on-board refrigeration
- Calibration stability lasts up to 90 days on most assays
Specifications
Each |
Safety and Handling
ShelfLife : 1 Year