Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience AIA-2000 Automated Immunoassay Analyzer
High-powered and flexible immunoassay testing.
Supplier: Tosoh Bioscience 022100
Description
- Automated daily maintenance
- Easy to load hybrid sorter
- Walk-away time ∽4 hours
- Two sampling arms
- Three incubators
- Two substrate bottles
- Load 960 tests (48 trays)
- 576 tips (6 tip racks)
- 200 tests per hour
- Volume: >1500 tests per month
- Dimensions: 907D x 1500W x 1260mm H (35.7 x 59.1 x 49.6 in.)
- Weight: 400kg (881.8 lb.)
- Large hospitals
- Reference labs
- Large clinics
- Core labs
Specifications
Automated Enzyme Immunoassay Analyzer | |
200 tests/hour | |
49.6 in. | |
59.1 in. | |
Top-to-top fluorescence detection | |
400 kg |
Large hospitals, Reference labs, Large clinics, Core labs | |
One-step and two-step sandwich and competitive FEIA | |
1260 mm | |
1500 mm | |
881.8 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction