Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tosoh Bioscience Waste Reservoir for AIA™-1800 Automated Immunoassay Analyzer
Accessory for use with AIA-1800 Automated Immunoassay Analyzer.
Supplier: Tosoh Bioscience 016320
Includes: Standard Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Total Laboratory Automation Model: CPU, monitor, printer, power conditioner (ABC PC 1200), UPS (1500), and accessory kit
Description
- Capacity: 10L
- 180 tests per hour with any combination AIA ST assays
Specifications
Water Reservoir | |
AIA-1800 automated immunoassay analyzer | |
10 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction