Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluidine Blue O, Fisher BioReagents™

Catalog Number BP10710
Click to view available options
Quantity:
10 g
Packaging:
Glass Bottle
92-31-9
C15H16ClN3S
305.82
MFCD00011934
HNONEKILPDHFOL-UHFFFAOYSA-M
7083
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
92-31-9
C15H16ClN3S
305.82
MFCD00011934
HNONEKILPDHFOL-UHFFFAOYSA-M
7083
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

This nuclear stain may be used as an alternative to methylene blue

TRUSTED_SUSTAINABILITY
Extinction Coefficient 700g(-1) cm(-1) min.
Merck Index 15, 9679
Solubility Information Soluble in water
Identification Pass Test
Lambda Max 620 to 638nm
Formula Weight 305.65
Loss on Drying 10% max.
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Color Index No. 52040
Color Green
Quantity 10 g
Physical Form Solid
Chemical Name or Material Toluidine Blue O
Show More Show Less
Recommended Storage RT
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit