Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Safe-Cote™
Steel Drum
Steel Pail
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.866g/cm³ |
Boiling Point | 111°C |
Quantity | 4 L |
Identification | Pass Test |
Packaging | Safe-Cote™ |
Physical Form | Liquid |
Assay Percent Range | ≥99.5 % |
Residue after Evaporation | 0.001% max. |
Show More |
Safety and Handling
Hazard Category | Signal Word: DANGER! |
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. NFPA Health: 2 Flammability: 3 Instability: 0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction