Learn More
Tocris Bioscience™ PHM 27 (human)
Potent calcitonin receptor agonist; endogenous analog of human VIP
Supplier:  Tocris Bioscience™ 2080/1
Catalog No. 20-801
Add to Cart
This item is not returnable. View return policy
Description
Description
PHM 27 (human) is an endogenous peptide product of human prepro-VIP and analog of porcine PHI-27; potent agonist for the human calcitonin receptor (EC50 = 11 nM). Transgenic mice expressing the human VIP/PHM 27 gene in pancreatic β-islets display enhanced glucose-induced insulin secretion.
Specifications
Specifications
1 mg