Learn More
MilliporeSigma™ Supelco™ TLC Saturation Pads
Pads improve reproducibility and reduce 'edge effects' when performing thin layer chromatography.
Supplier: MilliporeSigma™ Supelco™ Z2652411PAK
Description
Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor.Specifications
100/Pk. | |
Pk. of 100 Ea. | |
10 cm |
TLC | |
10 cm |