Learn More
Chemglass Life Sciences TLC Developing Tank Complete, Latch-Lid
Supplier: Chemglass Life Sciences CG118301
Heavy duty, molded TLC developing tank for use with plates up to 20 x 20 cm ( 8in x 8in). The weight of the tank is approximately 15 pounds and the bottom is ground to insure stability. The unique latching system holds the matching ground surfaces of the lid and tank firmly in place, enhancing the stability of the saturated atmosphere for the developing TLC plates. All of the metal components of the Latch-Lid mechanism are made of stainless steel. Inside dimensions of the tank are 27cm wide x 7cm deep x 26 cm high (10 5/8in x 2 7/8in x 10in). Tank is supplied complete with lid.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.