Learn More
Chemglass Life Sciences TLC Bottle only, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 8
Supplier: Chemglass Life Sciences CG118113
Replacement TLC bottle for the CG1181 series of TLC developing chambers. Bottle only, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 8.