Learn More
Cole-Parmer Titrating burette automatic, borosilicate glass; 50 mL
Supplier: Cole-Parmer 0604449
Constructed with a borosilicate glass 3.3 burette (buret). Flexible left- or right-handed operation possible. Accurately titrate with the micro-screw and discharge jet. Designed with a 50 mL capacity.