Learn More
Fine Science Tools TITANIUM BULLDOG SPRING CLAMP
Supplier: Fine Science Tools 1814430
Product: Titanium Bulldog Spring Clamp - Straight/30mm/ Shape: Straight/ Jaw Dimensions: 9.5 x 1mm/ Clamping Pressure: Light/ Alloy: Titanium/ Length: 30mm/ Serrated: Yes/ Unit: Each