Learn More
Costech Analytical Technologies Inc TIN CAPSULES 100/PK
Supplier: Costech Analytical Technologies Inc 041061
Tin capsules for solid samples,100/ box,Size: 5x9 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C A