Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticlopidine hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
53885-35-1
C14H15Cl2NS
300.24
MFCD00079606
MTKNGOHFNXIVOS-UHFFFAOYSA-N
ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf
65335
CHEBI:9589
hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
[H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2
This item is not returnable. View return policy
53885-35-1
C14H15Cl2NS
300.24
MFCD00079606
MTKNGOHFNXIVOS-UHFFFAOYSA-N
ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf
65335
CHEBI:9589
hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
[H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2

Selective P2Y12 antagonist

Selective P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity following oral administration in vivo.
Quantity 50 mg
Formula Weight 300.25
Percent Purity >99%
Chemical Name or Material Ticlopidine hydrochloride

Recommended Storage : Desiccate at Room Temperature