Learn More
Andwin Scientific TI-30X CALCULATOR
Supplier: Andwin Scientific Z12,3757
The Texas Instruments TI-30X IIS Scientific Calculator features 2-line display that exhibit the entry and the calculated result simultaneously. This product has three angle modes (degrees, radians, and grads) and scientific and engineering notation modes. It is recommended for general purpose use and for students who are enrolled in General Math, Pre-Algebra, Algebra 1 and 2, Geometry, Statistics, and General science.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.