Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Thumb Forceps PTFE Coated

Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
This item is not returnable.
View return policy
Description
Fisherbrand™ stainless steel forceps are PTFE-coated. Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
- Stainless steel construction
- Chemically resistant
- Nonsticking and non-corroding
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Lab Forceps | |
PTFE | |
Sample handling | |
Yes | |
4.5 in. | |
11.4 cm | |
No |
Yes | |
Stainless Steel | |
Forceps with PTFE coating | |
1 | |
4.5 in. | |
56 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title