Learn More
Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes
Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.
Manufacturer:  Wilmad-LabGlass™ 669995750
 View more versions of this product
Catalog No. 16-800-496
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications
Specifications
178 mm
ASTM E438 Type 1 Class B Glass
MHz Rating: High-Throughput
4.94665±0.01905 mm
0.43 mm
WG-1000-7-50
60 µm
NMR Tube
Disposable NMR Cap
7 in.
50/Pk.