Learn More
Applied Biosystems™ Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit
The Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit is designed to be used with fluorescent dye-labeled primers and high-resolution fluorescence scanners.
Manufacturer:  Applied Biosystems™ 792601KT
Encompass
Catalog No. 79-260-1KT
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Thermo Sequenase with thermostable inorganic Pyrophosphatase, Reaction Buffer (concentrate), Control DNA pUC19, Control Primer, TAMRA -40 Universal Forward Primer, ddG Termination Mix (7-deaza-dGTP), ddA Termination Mix (7-deaza-dGTP), ddT Termination Mix (7-deaza-dGTP), ddC Termination Mix (7-deaza-dGTP), Formamide Loading Dye, Protocol Book
Description
Description
Specifications
Specifications
Kit Components:
Thermo Sequenase with thermostable inorganic Pyrophosphatase
Reaction Buffer (concentrate)
Control DNA pUC19
Control Primer, TAMRA -40 Universal Forward Primer
ddG Termination Mix (7-deaza-dGTP)
ddA Termination Mix (7-deaza-dGTP)
ddT Termination Mix (7-deaza-dGTP)
ddC Termination Mix (7-deaza-dGTP)
Formamide Loading Dye
Protocol Book
The kit and all the enclosed reagents should be stored frozen at -20°C (NOT in a frost-free freezer). Keep all reagents on ice when removed from storage for use.
Sequencing
Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit
Kit
Universal Forward Primer
1 Kit