Learn More
Epredia™ Lab Vision™ UltraVision™ Quanto Mouse on Mouse HRP Detection System
HRP polymer designed to detect mouse antibodies on mouse tissue, maximizing signal while minimizing background.
$6066.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
TL125QHDM
|
Epredia™
TL125QHDM |
125 mL |
Each for $6,066.00
|
|
|||||
TL015QHDM
|
Epredia™
TL015QHDM |
15 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Epredia UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely.
UltraVision Quanto Makes Optimal Results Routine by providing:
- Strong Staining
- Reduced Mouse on Mouse Background
- Simple Biotin Free Polymer Procedure
- For use with a DAB or other HRP Chromogen; For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL)
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.