Learn More
Thermo Scientific™ Owl™ Separation B1A EasyCast™ Mini Gel Electrophoresis System
Designed to provide flat, even banding patterns and consistent results with less hassle.
Supplier: Thermo Scientific™ B1A/EMD
Description
The perfect size to determine molecular weight ranges of PCR fragments or larger-sized DNA molecules.
- Enables casting and running a gel in the buffer chamber
- Run 5 to 24 samples in 15 minutes
- The two comb slots on the UV Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
- Heavy-duty, one-piece combs eliminate comb assembly
- UVT gel trays are silk screened with a fluorescent ruler for easy measurement of bands
- 8L × 7cmW (3.1 × 2.8 in.) gel size
- 16L × 10.5W × 9.5cmH (6.3 × 4.1 × 3.7 in.) footprint
- 400mL running buffer volume
Specifications
EN61010-1-1993 (IEC1010-1), CE | |
Mini Gel Electrophoresis System | |
5 to 24 | |
400 mL | |
6.3 x 4.1 x 3.75 in. (16 x 10.5 x 9.5 cm) | |
3.1 in. | |
8 cm | |
2.7 in. | |
7 cm | |
9.5 cm |
30 to 60 min. | |
Main Buffer chamber, SuperSafe™ lid with attached power supply Leads, EasyCast™ gasketed UV transparent gel tray, two heavy duty, one piece, double sided combs: 6 and 10-well, 1/1.5mm thick | |
13.5 oz. | |
3 Years | |
6.3 in. | |
16 cm | |
4.1 in. | |
10.5 cm | |
3.75 in. | |
20/150 V |