Learn More
Thermo Scientific™ Orion Star™ A111 pH Benchtop Meter
Easy to use first time and every time, prepare students for laboratory and research applications using professionally proven benchtop meters.
Supplier: Thermo Scientific™ STARA1116/EMD
Includes: Orion Star™ A111 pH Benchtop Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter
Orion Star™ A111 pH Benchtop Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter
Orion Star™ A111 pH Benchtop Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter
Description
Thermo Scientific Orion Star A111 pH Benchtop Meters combine simplicity with accuracy. A large LCD displays pH or mV readings along with temperature. Icons provide quick updates on battery life, electrode status and calibration information. Simple button layout and onscreen messages help with calibration and setup menu choices.
- Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source
- Don't miss a reading — Auto-Read measurement mode locks in the stable reading on your screen and ready indicator alerts when readings are stable
- Up to three point pH calibration
- Automatic recognition of USA/NIST and DIN buffers
- Easily recall calibration slope data for procedure checks
- Non-volatile memory holds up to 50 data points
- Included electrode arm and newly-designed probe holder make it easier to maintain and place probes into samples
- Plug the meter into almost any power outlet with the included universal power adapter, or use four AA batteries (sold separately) to run the meter on DC power
- The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem
Specifications
- Accuracy (mV): ±0.2 mV or ±0.05% of reading whichever is greater
- Accuracy (pH): ±0.01pH
- Battery Life: 2000 hours
- Inputs: BNC (pH), 8-pin Mini-DIN (temperature)
- Probe Type: ±pH electrode, ATC probe, ORP electrode - purchase separately
- Range (mV): 1600.0mV
- Range (pH): -2.00 to 16.00pH
- Relative Humidity Range: 5 to 85 %, non-condensing
- Resolution (mV): 0.1mV
- Range (Relative mV): ±1999.9 RmV
- Resolution (pH): 0.1, 0.01pH
- Temperature (Metric) Operating: 5 to 45°C
- Temperature Range: -5° to +105°C
- Calibration Points (pH): 1 to 3
Certifications
CE, TÜV, FCC Class A limits , IP-54
Warranty and Services
Three-year meter replacement warranty
Specifications
pH, mV, Temperature | |
±1600 | |
-5°C to +105°C | |
Meter/Electrode |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.