Learn More
Thermo Scientific™ Orion™ 4-Star Plus pH/ISE Benchtop Multiparameter Meter
Measures pH and concentration with the accuracy and reliability you need for ISE measurements
Supplier: Thermo Scientific™ 1115003
Includes: Meter Only 1115000 package includes meter; universal power supply and instructions.
Meter Kit/pH 1115001 package includes meter; Ross™ Ultra pH electrode 8102BNUWP; epoxy ATC probe 927005MD; electrode stand; 1 pt. each of pH 4.01/7.00/10.01 buffers and storage solution; universal power supply and instructions.
Meter Kit/pH 1115002 package includes meter; SureFlow™ pH electrode 9172BNWP; epoxy ATC probe 927005MD; electrode stand; 1 pt. each of pH 4.01/7.00/10.01 buffers and storage solution; universal power supply and instructions.
Meter Kit/Sodium 1115005 package includes meter; Sodium ISE 8611BNWP; electrode stand; 1 pt. each of sodium standards and solutions; universal power supply and instructions.
Description
Improved calibration and measurement accuracy. Easy to set up and use while delivering exceptional, long-life performance for demanding R&D and QA/QC applications.
Easy to operate in any mode
- Bright, backlit LCD display with ready indication Simultaneous display of pH, mV, or ISE plus temperature
- Convenient built-in Quick-Start Guide with calibration instructions
- Stores, displays and prints up to 10 methods with individual method password protection
- Automatic or manual temperature compensation
- Auto-read/hold function
- Capable of operating an autosampler or stirrer (both sold separately)
- IP54 dustproof and splashproof
- pH: automatic or manual calibration; 1 to 5 calibration points; auto-buffer-recognition of NIST™/US and DIN buffers
- ISE: manual calibration; 1 to 5 calibration points; point-to-point calibration, high and low concentration range; automatic blank correction
- mV/ORP: EH autocalibration to standard hydrogen electrode
Increased accuracy
- Improved stability algorithm improves calibration and measurement accuracy
- SMART STABILITY™ and SMART AVERAGING™ functions optimize accuracy, precision and response time
- ATC calibration
- Advanced self-tests on meter operation, self-diagnostics
- Full complement of GLP-related features
- Optional Star Plus Navigator 21 software provides 21 CFR Part 11, GLP IQ/OQ documentation
- 1000 point datalogger, downloadable to printer or computer via RS-232 connection
- Average slope and slope of segment printed with measurement
Certifications
CE, CSA, UL, TUV and FCC Class A limits approved
Compliance
21 CFR Part II Compliant with optional software
Warranty and Services
Three years