Learn More
Thermo Scientific™ Open Top Caps Welded Septa
Click to view available options
Material:
Polypropylene
Description
- Polypropylene open top caps
- Welded PTFE/Silicone resealable septum
Order Info
Pre-cleaned caps are available by special request
Specifications
Material | Polypropylene |
Liner Material | PTFE-Faced Silicone Rubber |