Learn More
Thermo Scientific™ Thermo Scientific™ Nalgene™ Stainless Steel Autoclavable Dolly
For moving smalls tanks and carboys
Supplier: Thermo Scientific™ 26240020
Description
- Noncorrosive, made of stainless steel
- Casters do not leave marks on floor
Specifications
227.3 kg | |
20.9 in. | |
To move small tanks and carboys |
Stainless Steel | |
53 cm | |
Round |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.