Learn More
Thermo Scientific™ Matrix CNET 1500N Wired Data Router
A powerful device that uploads multi-port serial data to a TCP/IP or radio network
Supplier: Thermo Scientific™ SYS/1/70612/000
Description
The Matrix CNET-1500 N router is the standard “Serial to Ethernet” device with added features allowing any device to operate over a LAN network.
- Can run as a Wired Data Router, connecting directly into your 10/100Mbps Ethernet LAN, providing support for up to 4 RS232 devices, 255 RS485 devices, relays, and I/O control
- Can accept both incoming network connections and maintain outbound connections to remote monitoring software servers. This capability enables transparent serial tunnels to be created
- Can support data transmission both wired and wireless simultaneously if the optional wireless module is installed, providing redundant data availability for critical applications
- 10/100Base-T Ethernet
- 4 Serial Ports
- Easily interface legacy serial devices to Ethernet
- Extend the communication distance between 2 serial devices
- Metal enclosure for industrial applications
- Web browser interface for remote configuration
- Secure remote configuration
- AT command set emulation
- Application compatibility with Thermo Scientific ViewPoint and TeleTrak systems
Municipalities, chemical and nuclear plants, special events, critical facilities
Specifications
Wired Data Router | |
Municipalities, chemical and nuclear plants, special events, critical facilities |