Learn More
Thermo Scientific™ ABgene™ 2DCypher™ Cluster Rack Reader
Complete stand-alone unit for high-speed reading of 96 2DCypher-coded tubes in cluster rack format
Supplier: Thermo Scientific™ AB1066
Description
Features 3-second read time, rack orientation recognition, ability to integrate other scanning systems (rack barcode readers and single tube reader). ODBC-compatible results output; customizable result formats.
Components: Matrox 4Sight-II Unit PC with 850MHz Intel™ Pentium™ III processor, 10.4Gbytes storage, 128Mbytes RAM, standard 10/100BaseT LAN connection, preinstalled software. Reader head (207mm L × 120mm W × 170mm H) with integrated light source, temperature control down to -20°C, power supply. 15 in. TFT flat screen monitor. USB keyboard and mouse; 2 USB, 2 serial, 1 parallel (printer) ports, 3 main power leads. Reading head case safeguards against ingress of fluids and dust. T-Slot encasing permits mounting within integrated systems. Can be used in cold room storage. Compatible with some non-ABgene codes. Operating Parameters: Ambient temperature processor. -20°C to +30°C reading head. Power to ×3 IEC connections (100-240V at 50-60Hz).
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.