Learn More
Thermo Scientific THERMO CONCENTRIC NEBULIZER
Supplier: Thermo Scientific 842312051431
Glass Concentric Nebuliser; Standard concentric nebuliser for use with samples containing up to 3%; TDS.; Compatible with all iCAP 6000 Series Spectrometers.
This item is not returnable.
View return policy