Learn More
Texas Instruments™ ViewScreen™ Products for TI Calculators
Project the display of a graphing calculator for viewing by an entire class.
Supplier: Texas Instruments Inc 89TXSII/CBX
Includes: Both packages include: ViewScreen display panel, unit-to-unit link, AC adapter, guidebook, carrying case.
Description
- Using an overhead projector, image is bright enough to easily view, even with the lights on
- Students can display their own work quickly and easily
- Provides a more engaging, interactive classroom learning experience
- Saves time and provides more accurate results than blackboard diagrams
- 10 ft. cable links TI graphing calculators to ViewScreen panel
- ViewScreen with panel only works with calculators listed below.
- Package 1 is compatible with: TI-84 Plus Silver Edition TI-84 Plus, TI-83 Plus Silver Edition, TI-83 Plus, TI-83, TI-82, TI-73 Explorer™
Specifications
ViewScreen™ Package 3 | |
Project the display of a graphing calculator for viewing by an entire class | |
TI calculator | |
ViewScreen™ display type panel, unit to unit link, AC adapter, guidebook, carrying case |