Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrahydrofuran (Histological), Fisher Chemical™
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
Specifications
pH | 7 to 8 |
Melting Point | -108.4°C |
Color | Colorless |
Density | 0.880g/cm³ |
Boiling Point | 66°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Residue after Evaporation | Pass Test |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:2 Flammability:3 Instability:1 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction