Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroboric Acid Solution, Honeywell Fluka™

Catalog Number 6001665
Click to view available options
Quantity:
500 g
6 x 500 g
16872-11-0
BF4H
87.812
MFCD00011345
ODGCEQLVLXJUCC-UHFFFAOYSA-O
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
28118
CHEBI:38902
hydron;tetrafluoroborate
[H+].[B-](F)(F)(F)F
16872-11-0
BF4H
87.812
MFCD00011345
ODGCEQLVLXJUCC-UHFFFAOYSA-O
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
28118
CHEBI:38902
hydron;tetrafluoroborate
[H+].[B-](F)(F)(F)F

48 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Concentration 48wt. % in H2O
Density 1.4g/mL at 25°C
Vapor Pressure 5mmHg (20°C)
Quantity 6 x 500 g
Linear Formula HBF4
UN Number 1775
Formula Weight 87.81g/mol
Chemical Name or Material Tetrafluoroboric Acid Solution
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit