Learn More
Andwin Scientific TEMPILSTIK 250F/121C 10/PK
Supplier: Andwin Scientific 28019
Tempilstik provides accurate, precise, and convenient surface temperature measurement. This product is also ideal for use in pre-heating, interpass and post-weld heat treating applications. It is sold in a pack of 10.