Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Techne™ Hybridization Incubator
Temperature Range: Ambient +10°C to 100°C
Supplier: Techne™ 3945501
Includes: 3 large hybridization tubes FHB12
Description
Blotting techniques using radioactive and colorimetric probes are used in many areas of molecular biology. The expense and hazardous nature of these chemicals make it necessary that hybridization procedures should be safe, easy, use minimal volumes, and allow re-use of materials. Techne Hybridization Incubators provide the perfect solution and offer significant benefits over shaker bath/plastic bag methods.- Tubes and other accessories can be accommodated together for multiple uses
- Low probe volume, even with large glass tubes. Rotation and design ensure volumes as low as 5ml can be used and recovered
- Unique “slot-in” tubes; with tube rotation speed of 0 to 20 rpm; controllable to suit your application
- Radioactive safe; protective casing and non-drip tube design minimizes risk to the user
- Adjustable feet; for levelling on uneven surfaces
- Drip tray; removable for easy cleaning
- The Hybridizer HI-200 is easy to use and provides complete protection from hybridization hazards
- Temperature from 10°C above ambient to 100°C
- High capacity; can hold up to 24 mini tubes or 6 unique large tubes
- Hybridizations can be performed with a minimal volume of 5mL of probe
- Adjustable feet to enable accurate levelling
- Unique double-glazed glass door; quiet and safe, providing durable protection
Specifications
Ambient +10° to 100°C | |
Digital | |
4 rpm | |
±0.5°C | |
22.9 cm | |
10 in. | |
25.4 cm | |
14 in. | |
35.6 cm | |
15.2 cm | |
117 V |
Hybridization Incubator | |
HI-200 Hybridization incubator (120V), 3 large hybridization tubes FHB12 | |
4 rpm | |
9 in. | |
15 in. | |
38.1 cm | |
21 in. | |
53.3 cm | |
6 in. | |
117 V, 50/60 Hz | |
50/60 Hz |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction