Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ TECGEN™ FR Performance Utility Pants 32 in.
SDP

Catalog Number p-7163255
Click to view available options
Color:
Blue
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

National Safety Apparel™ TECGEN™ FR Performance Utility Pants with durable construction and high abrasion resistance.

  • CAT 2
  • Fabric/Material: 9.5 oz. High Performance FR Duck
  • Dual certified to protect against flash fire and arc flash hazards
  • Ultimate heavy duty performance utility pant
  • Durable construction with high abrasion resistance
  • Breathable fabric which increases moisture management
  • Cargo pocket on left side with flap
  • Double tool pocket on the right side
  • Fits variety of tools including Klein Pliers
  • Standard hip and back pockets
  • Made in the USA
Size X-Small
Material Denim
Color Blue
No. of Pockets 7
Arc Flash Rating 14 cal/cm2
Certifications/Compliance NFPA 70E for Arc Flash, UL Certified to NFPA 2112 for Flash Fire, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269
Size (English) Inseam 32 in.
Size (English) Waist 30 in.
For Use With (Application) Utility FR
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit