Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ TECGEN™ FR Performance Utility Pants 30 in.
Click to view available options
Color:
Blue
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Description
- CAT 2
- Fabric/Material: 9.5 oz. High Performance FR Duck
- Dual certified to protect against flash fire and arc flash hazards
- Ultimate heavy duty performance utility pant
- Durable construction with high abrasion resistance
- Breathable fabric which increases moisture management
- Cargo pocket on left side with flap
- Double tool pocket on the right side
- Fits variety of tools including Klein Pliers
- Standard hip and back pockets
- Made in the USA
Specifications
Size | X-Small |
Material | Denim |
Color | Blue |
No. of Pockets | 7 |
Arc Flash Rating | 14 cal/cm2 |
Certifications/Compliance | NFPA 70E for Arc Flash, UL Certified to NFPA 2112 for Flash Fire, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269 |
Size (English) Inseam | 30 in. |
Size (English) Waist | 30 in. |
For Use With (Application) | Utility FR |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction