Learn More
Phoenix Flow Systems Inc TDT CRISP CONTROL CELL 10/TEST
Supplier: Phoenix Flow Systems Inc CRTDP10
Control cells for the TdT antigen. CRISP control cells are stained along with your standard panel to provide a reliable, reproducible positive control.