Learn More
Cole-Parmer Digi-Sense Type-J High-Temperature Fiberglass-Insulated Probe Mini-Connector, Exposed 10ft 20 -Gauge
Supplier: Cole-Parmer 0846722
Fiberglass wire is not recommended for use in moist or wet environments. Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust.