Learn More
Cole-Parmer Compact PFA-Coated Thermocouple Probe, 4.5" L, Grounded d; Type J
Supplier: Cole-Parmer 0844110
General-purpose probes are designed for use with liquids, gases and semisolids. The response time is 2 seconds or greater. Probe comes with PVC handle. color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.