Learn More
General Data Healthcare™ SHUR/Mark™ HISTO/Print™ Data Management Super Software
With 2-D data matrix barcode printing capability and printer load balancing
Supplier: General Data Healthcare™ SMHPSS
Includes: One free license capable of operating one labeling device
Requires : Programming by institution's programmer or LIS vendor
Description
- Expansion of Advanced Software feature-set
- Accepts extrapolated ASCII formatted patient data from mainframes, networks and laboratory information systems
- Allows hand-free printing with Auto-Population feature
- Able to modify layouts with Drag and Drop Technology
- Allows PAUSE, PLAY, STOP and PREVIEW during actual print job
- Prints cassettes, slides, labels and other documents simultaneously without changing formats
- Adjustable printing speed and LIS Integration system
Specifications
Super Software |