Learn More
General Data Healthcare™ SHUR/Mark™ HISTO/Print™ Advanced Data Management Software
Allows customized printing of templates to enhance slide and cassette visibility and increase scan rates
Supplier: General Data Healthcare™ SMHPAS
Includes: One license for operating one labeler
Description
- Allows hand-free printing with Auto-Population feature
- Able to modify layouts with Drag and Drop Technology
- Prints barcodes in either dot or square formats
- Allows PAUSE, PLAY, STOP and PREVIEW during actual print job
- Comes with load balance feature to speed up batch printing processes
- Prints cassettes, slides, labels and other documents simultaneously without changing formats
- Adjustable printing speed and LIS Integration system
Specifications
Advanced Software |