Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Taurine, USP, 98-102%, Spectrum™ Chemical
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Taurine, USP is an organic acid found in animal tissues, specifically bile and the large intestine. Some of its roles include conjugation of bile acids, membrane stabilization and modulation of calcium signaling. It is crucial for cardiovascular function. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Ignition Residue | 0.003 |
Loss on Drying | 0.003 |
Packaging | Poly Pail |
Quantity | 5 kg |
Percent Purity | 98 to 102% |
Grade | USP |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction