Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tamsulosin Hydrochloride, EP, 98.5-101%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle

T1364, 106463-17-6, C20H28N2O5S•HCl

Spectrum™ Chemical Tamsulosin Hydrochloride, EP
CAS 106463-17-6
CAS Max % 1
Loss on Drying 0.005
Molecular Formula C20H29ClN2O5S
Packaging Amber Glass Bottle
Quantity 500 g
InChI Key ZZIZZTHXZRDOFM-XFULWGLBSA-N
SMILES [H+].[Cl-].CCOC1=CC=CC=C1OCCN[C@H](C)CC1=CC=C(OC)C(=C1)S(N)(=O)=O
IUPAC Name hydrogen 5-[(2R)-2-{[2-(2-ethoxyphenoxy)ethyl]amino}propyl]-2-methoxybenzene-1-sulfonamide chloride
Molecular Weight (g/mol) 444.97
Percent Purity 98.5 to 101%
Sulfated Ash 0.001
Grade EP
Show More Show Less