Learn More
Troemner™ Talboys™ Stainless Steel Erlenmeyer Flask Clamp
Manufactured to the highest standards. The Troemner™ Talboys™ Stainless Steel Erlenmeyer Flask Clamp keeps flasks secure.
Supplier: Troemner™ 30531475
Description
- Designed to hold Erlenmeyer flasks
- Constructed of type 302 and 304 stainless steel
- Includes hardware for easy attachment to platforms
- Flask clamps are supplied with a spring to hold flask in place