Learn More
Newcomer Supply Spirochete Treponema sp., Artificial Control Slides, 10/set
Supplier: Newcomer Supply 3746A
Newcomer Supply Spirochete Treponema sp., Artificial Control Slides are for the positive immunohistochemical staining of spirochetes, the causative agent of a variety of diseases such as; syphilis, bejel, pinta, yaws and lyme. Treponema hyodysenteriae purchased from American Type Culture Collection is used to produce the positive control tissue. Spirochete Treponema sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.